siRNA Details
| Gene Symbol: Gabrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183327.1 | NP_899156.1 | 9 | 1164 | ||
| XM_006246125.2 | XP_006246187.1 | 10 | 1751 | |||
| GenBank mRNA | AY574252.1 | - | 10 | 1188 | ||
| BC092617.1 | - | 5 | 451 | |||

Gabrg2
Genomic Map