siRNA Details
| Gene Symbol: Scn8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019266.2 | NP_062139.2 | 21 | 4261 | ||
| GenBank mRNA | AF049239.1 | - | 21 | 4261 | ||
| AF049240.1 | - | 21 | 4291 | |||
| L39018.1 | - | 21 | 4087 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn8a| Gene Symbol: Scn8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019266.2 | NP_062139.2 | 21 | 4261 | ||
| GenBank mRNA | AF049239.1 | - | 21 | 4261 | ||
| AF049240.1 | - | 21 | 4291 | |||
| L39018.1 | - | 21 | 4087 | |||