siRNA Details
| Gene Symbol: Kcnj5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017297.2 | NP_058993.1 | 2 | 708 | ||
| GenBank mRNA | BC087022.1 | - | 2 | 708 | 7192366 | |
| D50135.1 | - | 2 | 661 | |||
| L35771.1 | - | 2 | 713 | |||
| X83584.1 | - | 1 | 532 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj5| Gene Symbol: Kcnj5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017297.2 | NP_058993.1 | 2 | 708 | ||
| GenBank mRNA | BC087022.1 | - | 2 | 708 | 7192366 | |
| D50135.1 | - | 2 | 661 | |||
| L35771.1 | - | 2 | 713 | |||
| X83584.1 | - | 1 | 532 | |||