siRNA Details
Gene Symbol: Kcnj5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017297.2 | NP_058993.1 | 2 | 1083 | ||
GenBank mRNA | BC087022.1 | - | 2 | 1083 | 7192366 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcnj5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017297.2 | NP_058993.1 | 2 | 1083 | ||
GenBank mRNA | BC087022.1 | - | 2 | 1083 | 7192366 |