siRNA Details
| Gene Symbol: Cacna1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031601.3 | NP_113789.2 | 9 | 2703 | ||
| XM_008767985.1 | XP_008766207.1 | 9 | 2721 | |||
| XM_008767986.1 | XP_008766208.1 | 9 | 2721 | |||
| XM_008767987.1 | XP_008766209.1 | 9 | 2721 | |||
| XM_008767988.1 | XP_008766210.1 | 9 | 2721 | |||
| XM_008767989.1 | XP_008766211.1 | 9 | 2721 | |||
| XM_008767990.1 | XP_008766212.1 | 9 | 2721 | |||
| XM_008767991.1 | XP_008766213.1 | 9 | 2721 | |||
| XM_008767992.1 | XP_008766214.1 | 9 | 2721 | |||
| XM_008767993.1 | XP_008766215.1 | 9 | 2721 | |||
| XM_008767994.1 | XP_008766216.1 | 9 | 2721 | |||
| XM_008767995.1 | XP_008766217.1 | 9 | 2721 | |||
| XM_008767996.1 | XP_008766218.1 | 9 | 2721 | |||
| XM_008767997.1 | XP_008766219.1 | 9 | 2721 | |||
| XM_008767998.1 | XP_008766220.1 | 9 | 2721 | |||
| XM_008767999.1 | XP_008766221.1 | 9 | 2721 | |||
| XM_008768000.1 | XP_008766222.1 | 9 | 2670 | |||
| XM_008768001.1 | XP_008766223.1 | 9 | 2721 | |||
| GenBank mRNA | AF027984.1 | - | 9 | 2703 | ||
| AF125161.1 | - | 9 | 2378 | |||
| AF290212.2 | - | 9 | 2334 | |||
| EF116285.1 | - | 1 | 41 | |||
| EF116286.1 | - | 1 | 41 | |||

Cacna1g
Genomic Map