siRNA Details
| Gene Symbol: Kcnj16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053314.2 | NP_445766.2 | 1 | 880 | ||
| XM_006247568.2 | XP_006247630.1 | 2 | 1033 | |||
| XM_006247569.2 | XP_006247631.1 | 2 | 1120 | |||
| XM_006247570.2 | XP_006247632.1 | 2 | 1698 | |||
| XM_008768347.1 | XP_008766569.1 | 2 | 1124 | |||
| GenBank mRNA | BC078776.1 | - | 1 | 880 | 7107402 | |

Kcnj16
Genomic Map