siRNA Details
| Gene Symbol: Apbb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080478.1 | NP_536726.1 | 9 | 1419 | ||
| XM_006229935.1 | XP_006229997.1 | 9 | 1782 | |||
| XM_006229936.2 | XP_006229998.1 | 10 | 1624 | |||
| XM_006229938.2 | XP_006230000.1 | 10 | 1963 | |||
| XM_006229940.2 | XP_006230002.1 | 9 | 804 | |||
| XM_006229941.2 | XP_006230003.1 | 9 | 838 | |||
| XM_008759717.1 | XP_008757939.1 | 8 | 797 | |||
| GenBank mRNA | AF333983.1 | - | 9 | 1419 | ||
| X60469.1 | - | 9 | 1114 | |||

Apbb1
Genomic Map