siRNA Details
| Gene Symbol: Baat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017300.2 | NP_058996.2 | 4 | 1120 | ||
| GenBank mRNA | BC088153.1 | - | 4 | 1120 | 7368634 | |
| D43964.1 | - | 4 | 1135 | |||
| FQ218773.1 | - | 1164 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baat| Gene Symbol: Baat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017300.2 | NP_058996.2 | 4 | 1120 | ||
| GenBank mRNA | BC088153.1 | - | 4 | 1120 | 7368634 | |
| D43964.1 | - | 4 | 1135 | |||
| FQ218773.1 | - | 1164 | ||||