siRNA Details
| Gene Symbol: Slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017307.3 | NP_059003.1 | 2 | 247 | ||
| GenBank mRNA | BC100077.1 | - | 2 | 231 | 7126394 | |
| CB758644.1 | - | 2 | 263 | |||
| FQ214871.1 | - | 2 | 237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a1| Gene Symbol: Slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017307.3 | NP_059003.1 | 2 | 247 | ||
| GenBank mRNA | BC100077.1 | - | 2 | 231 | 7126394 | |
| CB758644.1 | - | 2 | 263 | |||
| FQ214871.1 | - | 2 | 237 | |||