siRNA Details
| Gene Symbol: Ywhae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031603.1 | NP_113791.1 | 2 | 321 | ||
| GenBank mRNA | BC063163.1 | - | 2 | 299 | 6921685 | |
| D30739.1 | - | 2 | 305 | |||
| FQ220393.1 | - | 2 | 327 | |||
| M84416.1 | - | 2 | 301 | |||
| U53882.1 | - | 2 | 321 | |||

Ywhae
Genomic Map