siRNA Details
| Gene Symbol: Colq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019274.2 | NP_062147.1 | 13, 14 | 1012 | ||
| GenBank mRNA | AF007583.1 | - | 13, 14 | 1012 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Colq| Gene Symbol: Colq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019274.2 | NP_062147.1 | 13, 14 | 1012 | ||
| GenBank mRNA | AF007583.1 | - | 13, 14 | 1012 | ||