siRNA Details
| Gene Symbol: Dpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105729.3 | NP_001099199.2 | 4, 5 | 494 | ||
| XM_006228677.2 | XP_006228739.1 | 4, 5 | 531 | |||
| XM_006228678.2 | XP_006228740.1 | 4, 5 | 528 | |||
| XM_006228679.2 | XP_006228741.1 | 4, 5 | 528 | |||
| XM_006228680.2 | XP_006228742.1 | 4, 5 | 533 | |||
| XM_006228681.2 | XP_006228743.1 | 4, 5 | 529 | |||
| XM_006228685.1 | XP_006228747.1 | 4, 5 | 1355 | |||
| XM_008759180.1 | XP_008757402.1 | 4, 5 | 517 | |||
| GenBank mRNA | BC159415.1 | - | 4, 5 | 422 | 8368092 | |
| CO396551.1 | - | 4, 5 | 494 | |||
| X66022.1 | - | 4, 5 | 1355 | |||

Dpf1
Genomic Map