siRNA Details
| Gene Symbol: Zfp292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008879.2 | NP_001008879.2 | 8 | 2736 | ||
| GenBank mRNA | L23077.1 | - | 1 | 1130 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp292| Gene Symbol: Zfp292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008879.2 | NP_001008879.2 | 8 | 2736 | ||
| GenBank mRNA | L23077.1 | - | 1 | 1130 | ||