siRNA Details
| Gene Symbol: Ugt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019276.3 | NP_062149.1 | 2 | 377 | ||
| GenBank mRNA | L21698.1 | - | 2 | 424 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt8| Gene Symbol: Ugt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019276.3 | NP_062149.1 | 2 | 377 | ||
| GenBank mRNA | L21698.1 | - | 2 | 424 | ||