siRNA Details
| Gene Symbol: Npepps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080395.1 | NP_536320.1 | 21 | 2643 | ||
| GenBank mRNA | FQ209839.1 | - | 16 | 1750 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npepps| Gene Symbol: Npepps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080395.1 | NP_536320.1 | 21 | 2643 | ||
| GenBank mRNA | FQ209839.1 | - | 16 | 1750 | ||