siRNA Details
Gene Symbol: Acot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031315.1 | NP_112605.1 | 3 | 886 | ||
GenBank mRNA | AB010428.1 | - | 3 | 881 | ||
Y09334.1 | - | 3 | 886 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
