siRNA Details
| Gene Symbol: Gjd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019281.2 | NP_062154.1 | 2 | 338 | ||
| GenBank mRNA | AJ296282.1 | - | 2 | 230 | ||
| Y16898.1 | - | 2 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gjd2| Gene Symbol: Gjd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019281.2 | NP_062154.1 | 2 | 338 | ||
| GenBank mRNA | AJ296282.1 | - | 2 | 230 | ||
| Y16898.1 | - | 2 | 338 | |||