siRNA Details
| Gene Symbol: Hspb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031607.1 | NP_113795.1 | 3 | 459 | ||
| GenBank mRNA | BC169050.1 | - | 3 | 447 | 7103781 | |
| CK358485.1 | - | 2 | 197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspb7| Gene Symbol: Hspb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031607.1 | NP_113795.1 | 3 | 459 | ||
| GenBank mRNA | BC169050.1 | - | 3 | 447 | 7103781 | |
| CK358485.1 | - | 2 | 197 | |||