siRNA Details
| Gene Symbol: Slc3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271089.2 | NP_001258018.1 | 1, 2 | 511 | ||
| NM_019283.3 | NP_062156.2 | 2, 3 | 636 | |||
| GenBank mRNA | AB015433.1 | - | 2, 3 | 602 | ||
| BC061989.1 | - | 1, 2 | 494 | 5599495 | ||
| FQ148223.1 | - | 1, 2 | 511 | |||
| U59324.1 | - | 1, 2 | 425 | |||

Slc3a2
Genomic Map