siRNA Details
| Gene Symbol: Cspg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019284.1 | NP_062157.1 | 2 | 1090 | ||
| NM_133652.1 | NP_598413.1 | 2 | 1078 | |||
| GenBank mRNA | AF292102.1 | - | 2 | 1078 | ||
| U33553.1 | - | 2 | 1090 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cspg5| Gene Symbol: Cspg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019284.1 | NP_062157.1 | 2 | 1090 | ||
| NM_133652.1 | NP_598413.1 | 2 | 1078 | |||
| GenBank mRNA | AF292102.1 | - | 2 | 1078 | ||
| U33553.1 | - | 2 | 1090 | |||