siRNA Details
Gene Symbol: Slco1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017111.1 | NP_058807.1 | 10 | 1265 | ||
GenBank mRNA | FQ209854.1 | - | 10 | 1283 | ||
L19031.1 | - | 10 | 1265 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slco1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017111.1 | NP_058807.1 | 10 | 1265 | ||
GenBank mRNA | FQ209854.1 | - | 10 | 1283 | ||
L19031.1 | - | 10 | 1265 |