siRNA Details
| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031610.3 | NP_113798.1 | 3 | 1073 | ||
| GenBank mRNA | L25264.1 | - | 3 | 1073 | ||
| U01071.1 | - | 3 | 1046 | |||
| U01141.1 | - | 3 | 1087 | |||
| U09243.1 | - | 1201 | ||||
| U60025.1 | - | 3 | 176 | |||
| U72410.1 | - | 4 | 618 | |||

Kcnj3
Genomic Map