siRNA Details
Gene Symbol: Aco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017321.1 | NP_059017.1 | 20 | 2756 | ||
GenBank mRNA | L23874.1 | - | 20 | 2756 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017321.1 | NP_059017.1 | 20 | 2756 | ||
GenBank mRNA | L23874.1 | - | 20 | 2756 |