siRNA Details
| Gene Symbol: Runx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017325.1 | NP_059021.1 | 4 | 947 | ||
| XM_008768598.1 | XP_008766820.1 | 4 | 2749 | |||
| GenBank mRNA | L35271.1 | - | 4 | 947 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001111023, NM_001111021, AK165307, D13802, CB056658, and others.

Runx1
Genomic Map