siRNA Details
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017327.1 | NP_059023.1 | 7 | 1482 | ||
| XM_008772320.1 | XP_008770542.1 | 7 | 1491 | |||
| XM_008772322.1 | XP_008770544.1 | 6 | 4100 | |||
| GenBank mRNA | AF413211.1 | - | 2 | 80 | ||
| AF413212.1 | - | 2 | 80 | |||
| M12671.1 | - | 6 | 646 | |||
| M17526.1 | - | 7 | 1482 | |||

Gnao1
Genomic Map