siRNA Details
| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017340.2 | NP_059036.1 | 789 | |||
| GenBank mRNA | BC085743.1 | - | 789 | 7109722 | ||
| J02752.1 | - | 772 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acox1| Gene Symbol: Acox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017340.2 | NP_059036.1 | 789 | |||
| GenBank mRNA | BC085743.1 | - | 789 | 7109722 | ||
| J02752.1 | - | 772 | ||||