siRNA Details
| Gene Symbol: Lipf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017341.2 | NP_059037.1 | 4 | 298 | ||
| GenBank mRNA | A01157.1 | - | 3 | 257 | ||
| X02309.1 | - | 3 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lipf| Gene Symbol: Lipf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017341.2 | NP_059037.1 | 4 | 298 | ||
| GenBank mRNA | A01157.1 | - | 3 | 257 | ||
| X02309.1 | - | 3 | 299 | |||