siRNA Details
| Gene Symbol: Slc6a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017348.2 | NP_059044.1 | 1 | 629 | ||
| XM_006229611.1 | XP_006229673.1 | 1 | 661 | |||
| GenBank mRNA | BC168238.1 | - | 1 | 607 | 5598758 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc6a8| Gene Symbol: Slc6a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017348.2 | NP_059044.1 | 1 | 629 | ||
| XM_006229611.1 | XP_006229673.1 | 1 | 661 | |||
| GenBank mRNA | BC168238.1 | - | 1 | 607 | 5598758 | |