siRNA Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017355.1 | NP_059051.1 | 7 | 641 | ||
| XM_008759181.1 | XP_008757403.1 | 6 | 860 | |||
| XM_008759182.1 | XP_008757404.1 | 6 | 860 | |||
| GenBank mRNA | FQ227674.1 | - | 7 | 673 | ||
| X78605.1 | - | 7 | 641 | |||

Rab4b
Genomic Map