siRNA Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017355.1 | NP_059051.1 | 6, 7 | 599 | ||
| XM_008759181.1 | XP_008757403.1 | 5, 6 | 818 | |||
| XM_008759182.1 | XP_008757404.1 | 5, 6 | 818 | |||
| GenBank mRNA | FQ227674.1 | - | 6, 7 | 631 | ||
| X78605.1 | - | 6, 7 | 599 | |||

Rab4b
Genomic Map