siRNA Details
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017361.1 | NP_059057.1 | 8 | 1122 | ||
GenBank mRNA | BC078858.1 | - | 8 | 1152 | 7134682 | |
U63840.1 | - | 8 | 1122 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nup54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017361.1 | NP_059057.1 | 8 | 1122 | ||
GenBank mRNA | BC078858.1 | - | 8 | 1152 | 7134682 | |
U63840.1 | - | 8 | 1122 |