siRNA Details
| Gene Symbol: Adcy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019285.2 | NP_062158.2 | 23 | 2900 | ||
| GenBank mRNA | BC081813.1 | - | 23 | 2900 | 7125413 | |
| FQ223268.1 | - | 22 | 3028 | |||
| FQ223451.1 | - | 22 | 3028 | |||
| M80633.1 | - | 24 | 2842 | |||

Adcy4
Genomic Map