siRNA Details
Gene Symbol: App | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019288.2 | NP_062161.1 | 7 | 1015 | ||
XM_006248008.2 | XP_006248070.1 | 7 | 1145 | |||
XM_006248009.2 | XP_006248071.1 | 7 | 1145 | |||
XM_006248010.2 | XP_006248072.1 | 7 | 1145 | |||
GenBank mRNA | AF513015.1 | - | 7 | 1016 | ||
BC062082.1 | - | 7 | 1128 | 5599880 | ||
FQ221425.1 | - | 7 | 1148 | |||
X14066.1 | - | 1 | 127 |
