siRNA Details
| Gene Symbol: Cdkn2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131902.3 | NP_571977.1 | 738 | |||
| GenBank mRNA | AF474980.2 | - | 259 | |||
| AF474981.2 | - | 240 | ||||
| BC088864.1 | - | 738 | 7319164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkn2c| Gene Symbol: Cdkn2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_131902.3 | NP_571977.1 | 738 | |||
| GenBank mRNA | AF474980.2 | - | 259 | |||
| AF474981.2 | - | 240 | ||||
| BC088864.1 | - | 738 | 7319164 | |||