siRNA Details
| Gene Symbol: Chrnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019297.1 | NP_062170.1 | 5 | 591 | ||
| GenBank mRNA | AY574258.1 | - | 5 | 403 | ||
| L31622.1 | - | 5 | 591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrnb2| Gene Symbol: Chrnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019297.1 | NP_062170.1 | 5 | 591 | ||
| GenBank mRNA | AY574258.1 | - | 5 | 403 | ||
| L31622.1 | - | 5 | 591 | |||