siRNA Details
| Gene Symbol: Chrnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019298.1 | NP_062171.1 | 10 | 1170 | ||
| GenBank mRNA | X74835.1 | - | 10 | 1170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrnd| Gene Symbol: Chrnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019298.1 | NP_062171.1 | 10 | 1170 | ||
| GenBank mRNA | X74835.1 | - | 10 | 1170 | ||