siRNA Details
| Gene Symbol: Cltc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019299.1 | NP_062172.1 | 27 | 4483 | ||
| XM_006247107.2 | XP_006247169.1 | 28 | 4795 | |||
| XM_006247108.2 | XP_006247170.1 | 27 | 4791 | |||
| XM_006247109.2 | XP_006247171.1 | 28 | 4794 | |||
| XM_008768108.1 | XP_008766330.1 | 26 | 5010 | |||
| GenBank mRNA | J03583.1 | - | 27 | 4483 | ||

Cltc
Genomic Map