siRNA Details
| Gene Symbol: Crk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019302.1 | NP_062175.1 | 2 | 304 | ||
| XM_006246913.1 | XP_006246975.1 | 2 | 482 | |||
| GenBank mRNA | D44481.1 | - | 2 | 304 | ||
| FQ210506.1 | - | 2 | 487 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crk| Gene Symbol: Crk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019302.1 | NP_062175.1 | 2 | 304 | ||
| XM_006246913.1 | XP_006246975.1 | 2 | 482 | |||
| GenBank mRNA | D44481.1 | - | 2 | 304 | ||
| FQ210506.1 | - | 2 | 487 | |||