siRNA Details
| Gene Symbol: Flt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019306.1 | NP_062179.1 | 16, 17 | 2593 | ||
| GenBank mRNA | D28498.1 | - | 16, 17 | 2593 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flt1| Gene Symbol: Flt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019306.1 | NP_062179.1 | 16, 17 | 2593 | ||
| GenBank mRNA | D28498.1 | - | 16, 17 | 2593 | ||