siRNA Details
| Gene Symbol: Gata4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144730.1 | NP_653331.1 | 5 | 1532 | ||
| XM_006252189.2 | XP_006252251.1 | 4 | 1112 | |||
| GenBank mRNA | L22761.1 | - | 5 | 1532 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gata4| Gene Symbol: Gata4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144730.1 | NP_653331.1 | 5 | 1532 | ||
| XM_006252189.2 | XP_006252251.1 | 4 | 1112 | |||
| GenBank mRNA | L22761.1 | - | 5 | 1532 | ||