siRNA Details
| Gene Symbol: Grik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019309.2 | NP_062182.1 | 10 | 1647 | ||
| GenBank mRNA | Z11548.1 | - | 1667 | |||
| Z11715.1 | - | 10 | 1445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grik2| Gene Symbol: Grik2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019309.2 | NP_062182.1 | 10 | 1647 | ||
| GenBank mRNA | Z11548.1 | - | 1667 | |||
| Z11715.1 | - | 10 | 1445 | |||