siRNA Details
| Gene Symbol: Cxcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019310.1 | NP_062183.1 | 1 | 276 | ||
| XM_006245273.2 | XP_006245335.2 | 2 | 353 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcr1| Gene Symbol: Cxcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019310.1 | NP_062183.1 | 1 | 276 | ||
| XM_006245273.2 | XP_006245335.2 | 2 | 353 | |||