siRNA Details
Gene Symbol: Cxcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019310.1 | NP_062183.1 | 1 | 143 | ||
XM_006245273.2 | XP_006245335.2 | 2 | 220 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cxcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019310.1 | NP_062183.1 | 1 | 143 | ||
XM_006245273.2 | XP_006245335.2 | 2 | 220 |