siRNA Details
| Gene Symbol: Kcnn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019314.1 | NP_062187.1 | 4 | 1053 | ||
| XM_006254682.2 | XP_006254744.1 | 4 | 2606 | |||
| XM_006254683.2 | XP_006254745.1 | 5 | 3679 | |||
| GenBank mRNA | DQ137427.1 | - | 2 | 281 | ||
| JN857942.1 | - | 4 | 1053 | |||
| U69882.1 | - | 4 | 1053 | |||

Kcnn2
Genomic Map