siRNA Details
| Gene Symbol: Mcpt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019323.1 | NP_062196.1 | 523 | |||
| GenBank mRNA | U67912.1 | - | 504 | |||
| U72143.1 | - | 523 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcpt9| Gene Symbol: Mcpt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019323.1 | NP_062196.1 | 523 | |||
| GenBank mRNA | U67912.1 | - | 504 | |||
| U72143.1 | - | 523 | ||||