siRNA Details
| Gene Symbol: Pfkfb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019333.1 | NP_062206.1 | 4 | 281 | ||
| XM_006243840.2 | XP_006243902.1 | 3 | 240 | |||
| XM_006243841.2 | XP_006243903.1 | 3 | 234 | |||
| XM_006243842.2 | XP_006243904.1 | 3 | 202 | |||
| XM_008766563.1 | XP_008764785.1 | 3 | 1281 | |||
| GenBank mRNA | EU404101.1 | - | 3 | 250 | ||
| EU404102.1 | - | 3 | 250 | |||
| EU404103.1 | - | 3 | 250 | |||
| M64797.1 | - | 4 | 281 | |||

Pfkfb4
Genomic Map