siRNA Details
| Gene Symbol: Pitx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042505.1 | NP_001035970.1 | 3 | 894 | ||
| NM_019334.2 | NP_062207.1 | 4 | 595 | |||
| GenBank mRNA | AF039832.1 | - | 4 | 595 | ||
| AJ222971.1 | - | 3 | 902 | |||
| AJ222972.1 | - | 4 | 648 | |||
| EF519321.1 | - | 3 | 384 | |||

Pitx2
Genomic Map