siRNA Details
| Gene Symbol: Rgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019336.1 | NP_062209.1 | 4, 5 | 496 | ||
| XM_006249974.2 | XP_006250036.1 | 4, 5 | 680 | |||
| GenBank mRNA | BC098681.1 | - | 4, 5 | 496 | 7385088 | |
| FQ234088.1 | - | 4, 5 | 394 | |||
| U77698.1 | - | 1, 2 | 171 | |||

Rgs1
Genomic Map