siRNA Details
| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019338.1 | NP_062211.1 | 798 | |||
| GenBank mRNA | CB605941.1 | - | 686 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs11| Gene Symbol: Rgs11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019338.1 | NP_062211.1 | 798 | |||
| GenBank mRNA | CB605941.1 | - | 686 | |||