siRNA Details
| Gene Symbol: Rgs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019341.1 | NP_062214.1 | 4 | 289 | ||
| GenBank mRNA | AF241259.1 | - | 4 | 289 | ||
| EF568934.1 | - | 4 | 353 | |||
| FQ234716.1 | - | 4 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rgs5| Gene Symbol: Rgs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019341.1 | NP_062214.1 | 4 | 289 | ||
| GenBank mRNA | AF241259.1 | - | 4 | 289 | ||
| EF568934.1 | - | 4 | 353 | |||
| FQ234716.1 | - | 4 | 406 | |||